(R)-1-Boc-3-Cyanopyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-tert-Butyl 3-cyanopyrrolidine-1-carboxylate

073628-1G 073628-5G
102531-172EA 238.46 USD
102531-172 102531-174
(R)-1-Boc-3-Cyanopyrrolidine ≥95%
(R)-1-Boc-3-Cyanopyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂
MW: 196.2500 g/mol
MDL Number: MFCD05863904
CAS Number: 132945-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR