Morpholin-2-ylmethanol ≥95%

Supplier: Matrix Scientific

073626-250MG 073626-1G 073626-5G
102531-160EA 250.77 USD
102531-160 102531-162 102531-164
Morpholin-2-ylmethanol ≥95%
Morpholin-2-ylmethanol

Hazard: Irritant

Formula: C₅H₁₁NO₂
MDL Number: MFCD03426271
CAS Number: 103003-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR