4-Bromo-2-benzofuran-1[3H]-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromophthalide

073625-250MG 073625-1G 073625-5G
102531-154EA 284.76 USD
102531-154 102531-156 102531-158
4-Bromo-2-benzofuran-1[3H]-one ≥95%
4-Bromo-2-benzofuran-1[3H]-one

Hazard: Irritant

Formula: C₈H₅BrO₂
MDL Number: MFCD06412577
CAS Number: 102308-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR