3-Fluoropyrrolidine ≥95%

Supplier: Matrix Scientific

073624-250MG 073624-1G 073624-5G
102531-148EA 569.23 USD
102531-148 102531-150 102531-152
3-Fluoropyrrolidine ≥95%
3-Fluoropyrrolidine

Hazard: Irritant

Formula: C₄H₈FN
MDL Number: MFCD06738454
CAS Number: 116574-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR