2,6-Dibromo-5-ethoxypyridin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-2,6-dibromo-5-ethoxypyridine

073623-250MG 073623-1G 073623-5G
102531-142EA 456.92 USD
102531-142 102531-144 102531-146
2,6-Dibromo-5-ethoxypyridin-3-amine ≥95%
2,6-Dibromo-5-ethoxypyridin-3-amine

Hazard: Irritant

Formula: C₇H₈Br₂N₂O
CAS Number: 1000018-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR