2-Bromonicotinamide ≥95%

Supplier: Matrix Scientific

073617-1G 073617-5G 073617-10G
102531-110EA 307.69 USD
102531-110 102531-112 102531-114
2-Bromonicotinamide ≥95%
2-Bromonicotinamide

Hazard: Irritant

Formula: C₆H₅BrN₂O
MW: 201.0207 g/mol
MDL Number: MFCD00234155
CAS Number: 87674-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR