(2-Chlorophenyl)acetylene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-2-ethynylbenzene

073616-1G 073616-5G 073616-10G
102531-104EA 30.77 USD
102531-104 102531-106 102531-108
(2-Chlorophenyl)acetylene ≥95%
(2-Chlorophenyl)acetylene

Hazard: Irritant/Light Sensitive/Refrigerate/Stored under Argon

Formula: C₈H₅Cl
MW: 136.58 g/mol
Boiling Pt: 82 °C (12 mmHg)
Density: 1.125
Flash Pt: 70 °C
MDL Number: MFCD00269951
CAS Number: 873-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR