(S)-4-Boc-morpholine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (2S)-Morpholine-2-carboxylic acid, N-BOC protected, (S)-4-Boc-Morpholine-2-carboxylic acid

073614-250MG 073614-1G
102531-096EA 684.62 USD
102531-096 102531-098
(S)-4-Boc-morpholine-2-carboxylic acid ≥95%
(S)-4-Boc-morpholine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MW: 231.2457 g/mol
MDL Number: MFCD06200662
CAS Number: 868689-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR