4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole ≥95%
Supplier: Matrix Scientific
073612-1G
073612-5G
102531-088EA
533.85
USD
102531-088
102531-090
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole ≥95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-trityl-1H-pyrazole
Hazard: Irritant
Formula:
C₂₈H₂₉BN₂O₂ MW: 436.3531 g/mol |
MDL Number:
MFCD10698507 CAS Number: 863238-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...