(2-Fluoropyridin-3-yl)methanamine ≥95%

Supplier: Matrix Scientific

073611-250MG 073611-1G
102531-082EA 487.24 USD
102531-082 102531-084
(2-Fluoropyridin-3-yl)methanamine ≥95%
(2-Fluoropyridin-3-yl)methanamine

Hazard: Irritant

Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD06213866
CAS Number: 205744-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR