N1,N2-Dimethylcyclohexane-1,2-diamine ≥95%
Supplier: Matrix Scientific
073610-5G
073610-25G
102531-078EA
198.46
USD
102531-078
102833-646
N1,N2-Dimethylcyclohexane-1,2-diamine ≥95%
N1,N2-Dimethylcyclohexane-1,2-diamine
Hazard: Irritant
Formula:
C₈H₁₈N₂ MW: 142.2419 g/mol |
MDL Number:
MFCD05664382 CAS Number: 61798-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...