4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane ≥95%

Supplier: Matrix Scientific

Synonyms: 8-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,4-dioxaspiro[4.5]dec-7-ene, 4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane, 1,4-Dioxaspiro[4,5]dec-7-en-8-boronic acid Pinacol ester

073609-1G 073609-5G
102531-072EA 136.92 USD
102531-072 102531-074
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane ≥95%
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1,3,2-dioxaborolane

Hazard: Irritant/Refrigerate

Formula: C₁₄H₂₃BO₄
MW: 266.1410 g/mol
MDL Number: MFCD04115656
CAS Number: 680596-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR