6-Bromo-1-tetralone ≥95%

Supplier: Matrix Scientific

073606-250MG 073606-1G 073606-5G
102531-054EA 301 USD
102531-054 102531-056 102531-058
6-Bromo-1-tetralone ≥95%
6-Bromo-1-tetralone

Hazard: Irritant

Formula: C₁₀H₉BrO
MDL Number: MFCD04114378
CAS Number: 66361-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR