7-Fluoro-2,3-dihydroinden-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Fluoroindan-1-one

073605-1G 073605-5G
102531-050EA 640 USD
102531-050 102531-052
7-Fluoro-2,3-dihydroinden-1-one ≥95%
7-Fluoro-2,3-dihydroinden-1-one

Hazard: Irritant

Formula: C₉H₇FO
MW: 150.1500 g/mol
MDL Number: MFCD07368749
CAS Number: 651735-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR