4-Chloro-2-methylquinazoline ≥95%

Supplier: Matrix Scientific

073604-1G 073604-5G
102531-046EA 121.54 USD
102531-046 102531-048
4-Chloro-2-methylquinazoline ≥95%
4-Chloro-2-methylquinazoline

Hazard: Irritant

Formula: C₉H₇ClN₂
MDL Number: MFCD05666292
CAS Number: 6484-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR