Ethyl-5-chloro-1,3,4-thiadiazole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-chloro-1,3,4-thiadiazole-2-carboxylate

073603-1G 073603-5G
102531-042EA 769.23 USD
102531-042 102531-044
Ethyl-5-chloro-1,3,4-thiadiazole-2-carboxylate ≥95%
Ethyl-5-chloro-1,3,4-thiadiazole-2-carboxylate

Hazard: Irritant

Formula: C₅H₅ClN₂O₂S
MW: 192.6234 g/mol
MDL Number: MFCD11112114
CAS Number: 64837-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR