4-Quinolinol ≥95%

Supplier: Matrix Scientific

073599-5G 073599-100G 073599-25G
102531-018EA 41.54 USD
102531-018 102531-022 102833-642
4-Quinolinol ≥95%
4-Quinolinol

Hazard: Irritant

Formula: C₉H₇NO
Melting Pt: 201…205 °C
MDL Number: MFCD00006777
CAS Number: 611-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR