5-Nitrouracil ≥95%

Supplier: Matrix Scientific

073598-10G 073598-100G
102531-014EA 72.31 USD
102531-014 102531-016
5-Nitrouracil ≥95%
5-Nitrouracil

Hazard: Irritant

Formula: C₄H₃N₃O₄
MDL Number: MFCD00006021
CAS Number: 611-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR