3-Amino-1-benzylpiperidine ≥95%

Supplier: Matrix Scientific

073597-1G 073597-5G 073597-10G
102531-008EA 81.54 USD
102531-008 102531-010 102531-012
3-Amino-1-benzylpiperidine ≥95%
3-Amino-1-benzylpiperidine

Hazard: Irritant

Formula: C₁₂H₁₈N₂
Boiling Pt: 349 °C
Flash Pt: 164 °C
MDL Number: MFCD04038444
CAS Number: 60407-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR