(S)-2-Methylpyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

073596-1G 073596-5G
102531-002EA 84.62 USD
102531-002 102531-004
(S)-2-Methylpyrrolidine hydrochloride ≥95%
(S)-2-Methylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClN
MW: 121.6100 g/mol
MDL Number: MFCD06796611
CAS Number: 174500-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR