2,3-Dihydroquinolin-4(1H)-one hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2,3,4-Tetrahydro-4-quinolinone hydrochloride

073594-1G 073594-5G
102530-994EA 569.23 USD
102530-994 102530-996
2,3-Dihydroquinolin-4(1H)-one hydrochloride ≥95%
2,3-Dihydroquinolin-4(1H)-one hydrochloride

Hazard: Irritant

Formula: C₉H₁₀ClNO
MW: 183.6400 g/mol
MDL Number: MFCD11052533
CAS Number: 71412-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR