5-Bromo-4-methylpyridine-2,3-diamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2,3-diamino-4-methylpyridine

073592-250MG 073592-1G 073592-5G
102530-984EA 400 USD
102530-984 102530-986 102530-988
5-Bromo-4-methylpyridine-2,3-diamine ≥95%
5-Bromo-4-methylpyridine-2,3-diamine

Hazard: Irritant

Formula: C₆H₈BrN₃
MW: 202.0518 g/mol
MDL Number: MFCD08062974
CAS Number: 41230-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR