6-Chloro-2-methoxy-3-nitropyridine ≥95%
Supplier: Matrix Scientific
073591-1G
073591-5G
102530-980EA
640
USD
102530-980
102530-982
6-Chloro-2-methoxy-3-nitropyridine ≥95%
6-Chloro-2-methoxy-3-nitropyridine
Hazard: Irritant
Formula:
C₆H₅ClN₂O₃ MW: 188.5700 g/mol |
MDL Number:
MFCD09033798 CAS Number: 40851-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...