2-Chloro-5-fluoro-3-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-fluoro-3-picoline

073588-1G 073588-5G
102530-966EA 49.23 USD
102530-966 102530-968
2-Chloro-5-fluoro-3-methylpyridine ≥95%
2-Chloro-5-fluoro-3-methylpyridine

Hazard: Irritant

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD04972725
CAS Number: 38186-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR