5,7-Difluoroquinazolin-4(3H)-one ≥95%

Supplier: Matrix Scientific

073587-1G 073587-5G
102530-962EA 458.46 USD
102530-962 102530-964
5,7-Difluoroquinazolin-4(3H)-one ≥95%
5,7-Difluoroquinazolin-4(3H)-one

Hazard: Irritant

Formula: C₈H₄F₂N₂O
MDL Number: MFCD11100783
CAS Number: 379228-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR