Ethyl-4-chloronicotinate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-chloronicotinate hydrochloride

073586-1G 073586-5G 073586-10G
102530-954EA 640 USD
102530-954 102530-956 102530-958
Ethyl-4-chloronicotinate hydrochloride ≥95%
Ethyl-4-chloronicotinate hydrochloride

Hazard: Irritant

Formula: C₈H₉Cl₂NO₂
MW: 222.700 g/mol
MDL Number: MFCD06804523
CAS Number: 174496-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR