Ethyl-6-chloroindole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-chloroindole-2-carboxylate

073585-5G 073585-25G
102530-950EA 104.62 USD
102530-950 102833-640
Ethyl-6-chloroindole-2-carboxylate ≥95%
Ethyl-6-chloroindole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₂
MDL Number: MFCD03084731
CAS Number: 27034-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR