4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

073583-1G 073583-5G
102530-940EA 640 USD
102530-940 102530-942
4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride ≥95%
4,5,6,7-Tetrahydro-1H-benzo[d]imidazole-5-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₈H₁₁ClN₂O₂
MW: 202.6381 g/mol
MDL Number: MFCD18157694
CAS Number: 131020-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR