4-Bromo-3-fluorobenzenesulfonamide ≥95%

Supplier: Matrix Scientific

073582-1G 073582-5G 073582-10G
102530-934EA 38.46 USD
102530-934 102530-936 102530-938
4-Bromo-3-fluorobenzenesulfonamide ≥95%
4-Bromo-3-fluorobenzenesulphonamide

Hazard: Irritant

Formula: C₆H₅BrFNO₂S
MW: 254.08 g/mol
MDL Number: MFCD03094292
CAS Number: 263349-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR