2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile ≥95%

Supplier: Matrix Scientific

073579-250MG 073579-1G 073579-5G
102530-918EA 281.52 USD
102530-918 102530-920 102530-922
2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile ≥95%
2,3-Dihydro-1-oxo-1H-indene-5-carbonitrile

Hazard: Irritant

Formula: C₁₀H₇NO
MW: 157.1700 g/mol
Melting Pt: 128…130 °C
MDL Number: MFCD08234685
CAS Number: 25724-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR