(1H-Pyrazol-4-yl)methanol ≥95%

Supplier: Matrix Scientific

073577-1G 073577-5G
102530-908EA 152.31 USD
102530-908 102530-910
(1H-Pyrazol-4-yl)methanol ≥95%
(1H-Pyrazol-4-yl)methanol

Hazard: Irritant

Formula: C₄H₆N₂O
MDL Number: MFCD06797673
CAS Number: 25222-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR