(R)-(+)-3-Hydroxypiperidine hydrochloride ≥95%
Supplier: Matrix Scientific
073576-1G
073576-5G
073576-10G
102530-900EA
355.38
USD
102530-900
102530-902
102530-904
(R)-(+)-3-Hydroxypiperidine hydrochloride ≥95%
(R)-(+)-3-Hydroxypiperidine hydrochloride
Hazard: Irritant
Formula:
C₅H₁₂ClNO |
MDL Number:
MFCD00192221 CAS Number: 198976-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...