(R)-(+)-1-Boc-3-methylpiperazine ≥95%

Supplier: Matrix Scientific

Synonyms: (3R)-3-Methylpiperazine, N1-BOC protected, (R)-1-Boc-3-Methylpiperazine, (R)-1-Boc-3-methyl-piperazine

073575-1G 073575-5G 073575-10G
102530-894EA 146.15 USD
102530-894 102530-896 102530-898
(R)-(+)-1-Boc-3-methylpiperazine ≥95%
(R)-(+)-1-Boc-3-methylpiperazine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD02683205
CAS Number: 163765-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR