2-Cyano-6-methylpyridine ≥95%
Supplier: Matrix Scientific
073574-5G
073574-25G
102530-890EA
29.23
USD
102530-890
102833-636
2-Cyano-6-methylpyridine ≥95%
2-Cyano-6-methylpyridine
Hazard: Irritant
Formula:
C₇H₆N₂ Melting Pt: 70…74 °C |
MDL Number:
MFCD00190586 CAS Number: 1620-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...