2-Cyanoisonicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-4-pyridinecarboxylic acid

073573-1G 073573-5G
102530-884EA 276.92 USD
102530-884 102530-886
2-Cyanoisonicotinic acid ≥95%
2-Cyanoisonicotinic acid

Hazard: Irritant

Formula: C₇H₄N₂O₂
MW: 148.1200 g/mol
MDL Number: MFCD03411729
CAS Number: 161233-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR