6-Bromo-3,4-dihydro-2H-isoquinolin-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-3,4-dihydroisoquinolin-1(2H)-one

073571-1G 073571-5G
102530-872EA 246.15 USD
102530-872 102530-874
6-Bromo-3,4-dihydro-2H-isoquinolin-1-one ≥95%
6-Bromo-3,4-dihydro-2H-isoquinolin-1-one

Hazard: Irritant

Formula: C₉H₈BrNO
MW: 226.0700 g/mol
MDL Number: MFCD04114858
CAS Number: 147497-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR