N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide ≥95%
Supplier: Matrix Scientific
Synonyms:
(1R,2R)-(-)-N-(4-toluenesulphonyl)-1,2-diphenylethylenediamine, (R,R)-TsDPEN, (1R,2R)-(-)-N-(4-Toluenesulphonyl)-1,2-diphenylethylenediamine
073569-5G
073569-25G
102530-862EA
167.69
USD
102530-862
102551-850
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide ≥95%
N-((1R,2R)-2-Amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide
Hazard: Irritant
Formula:
C₂₁H₂₂N₂O₂S MW: 366.48 g/mol Melting Pt: 125 °C |
MDL Number:
MFCD02093428 CAS Number: 144222-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...