Methyl indoline-5-carboxylate ≥95%

Supplier: Matrix Scientific

073567-1G 073567-5G
102530-852EA 106.15 USD
102530-852 102530-854
Methyl indoline-5-carboxylate ≥95%
Methyl indoline-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.1998 g/mol
MDL Number: MFCD07371630
CAS Number: 141452-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR