(S)-3-(Aminomethyl)-1-boc-piperidine ≥95%

Supplier: Matrix Scientific

073564-1G 073564-5G 073564-10G
102530-836EA 284.62 USD
102530-836 102530-838 102530-840
(S)-3-(Aminomethyl)-1-boc-piperidine ≥95%
(S)-3-(Aminomethyl)-1-Boc-piperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD03839877
CAS Number: 140645-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR