(S)-1-Boc-pyrrolidine-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-1-Boc-Pyrrolidine-3-carboxylic acid, (3S)-1-tert-butoxycarbonylpyrrolidine-3-carboxylic acid

073563-1G 073563-5G
102530-832EA 153.85 USD
102530-832 102530-834
(S)-1-Boc-pyrrolidine-3-carboxylic acid ≥95%
(S)-1-Boc-pyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD03094728
CAS Number: 140148-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR