1-Phenyl-1H-pyrazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

073561-250MG 073561-1G
102530-820EA 456.92 USD
102530-820 102530-822
1-Phenyl-1H-pyrazole-5-carbaldehyde ≥95%
1-Phenyl-1H-pyrazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₈N₂O
MW: 172.1800 g/mol
MDL Number: MFCD00103497
CAS Number: 132274-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR