2-Chloro-4,6-dimethoxypyrimidine ≥95%
Supplier: Matrix Scientific
073560-10G
073560-100G
102530-816EA
126.15
USD
102530-816
102530-818
2-Chloro-4,6-dimethoxypyrimidine ≥95%
2-Chloro-4,6-dimethoxypyrimidine
Hazard: Irritant
Formula:
C₆H₇ClN₂O₂ MW: 174.58 g/mol |
MDL Number:
MFCD00274530 CAS Number: 13223-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...