4-Bromo-2-formylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromopicolinaldehyde, 4-Bromopyridine-2-carbaldehyde

073559-1G 073559-5G
102530-810EA 138.46 USD
102530-810 102530-812
4-Bromo-2-formylpyridine ≥95%
4-Bromo-2-formylpyridine

Hazard: Irritant/Refrigerate

Formula: C₆H₄BrNO
MW: 186.01 g/mol
MDL Number: MFCD08690697
CAS Number: 131747-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR