Benzo[b]thiophen-3(2H)-one ≥95%

Supplier: Matrix Scientific

073557-250MG 073557-1G 073557-5G
102530-798EA 498.46 USD
102530-798 102530-800 102530-802
Benzo[b]thiophen-3(2H)-one ≥95%
Benzo[b]thiophen-3(2H)-one

Hazard: Irritant

Formula: C₈H₆OS
MDL Number: MFCD08061607
CAS Number: 130-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR