2-(2-Bromophenyl)pyrrolidine ≥95%

Supplier: Matrix Scientific

073555-1G 073555-5G
102530-788EA 129.46 USD
102530-788 102530-790
2-(2-Bromophenyl)pyrrolidine ≥95%
2-(2-Bromophenyl)pyrrolidine

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₂BrN
MW: 226.1130 g/mol
MDL Number: MFCD02663433
CAS Number: 129540-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR