4-Amino-6-methylnicotinic acid ≥95%

Supplier: Matrix Scientific

073553-250MG 073553-1G 073553-5G
102530-778EA 250.11 USD
102530-778 102530-780 102530-782
4-Amino-6-methylnicotinic acid ≥95%
4-Amino-6-methylnicotinic acid

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.1506 g/mol
MDL Number: MFCD12923129
CAS Number: 127915-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR