4-Bromo-2-fluorophenylacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Bromo-2-fluorophenyl)acetic acid

073552-1G 073552-5G
102530-772EA 93.85 USD
102530-772 102530-774
4-Bromo-2-fluorophenylacetic acid ≥95%
4-Bromo-2-fluorophenylacetic acid

Hazard: Irritant

Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
MDL Number: MFCD09032952
CAS Number: 114897-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR