Benzo[d]thiazol-7-amine ≥95%

Supplier: Matrix Scientific

073551-1G 073551-5G
102530-768EA 415.38 USD
102530-768 102530-770
Benzo[d]thiazol-7-amine ≥95%
Benzo[d]thiazol-7-amine

Hazard: Irritant

Formula: C₇H₆N₂S
MDL Number: MFCD10566715
CAS Number: 1123-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR