2-Methylpyridin-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-2-methylpyridine

073550-5G 073550-25G
102530-762EA 43.08 USD
102530-762 102551-848
2-Methylpyridin-3-ol ≥95%
2-Methylpyridin-3-ol

Hazard: Irritant

Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 166-169°C
MDL Number: MFCD00082538
CAS Number: 1121-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR