4-Methylpyridin-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-4-methylpyridine

073549-1G 073549-5G
102530-756EA 158.46 USD
102530-756 102530-758
4-Methylpyridin-3-ol ≥95%
4-Methylpyridin-3-ol

Hazard: Irritant

Formula: C₆H₇NO
MW: 109.1259 g/mol
MDL Number: MFCD04114227
CAS Number: 1121-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR